missing translation for 'onlineSavingsMsg'
Learn More

MG-101, MedChemExpress

Product Code. 16497658
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497658 10 mg 10mg
16493667 50 mg 50mg
16460128 5 mg 5mg
16427566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497658 Supplier Medchem Express Supplier No. HY18964
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MG-101 (ALLN) is an inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. MG-101 induces apoptosis and inhibits tumor growth, it can be used for the research of colon cancer.

MG-101 (ALLN) is an inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. MG-101 induces apoptosis and inhibits tumor growth, it can be used for the research of colon cancer.

Specifications

Chemical Name or Material MG-101
Physical Form Solid
CAS 110044-82-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H37N3O4
For Use With (Application) Cancer-Kinase/protease
Synonym Calpain inhibitor I Ac-LLnL-CHO ALLN
Solubility Information DMSO : 16.67 mg/mL (43.46 mM; Need ultrasonic)
SMILES O=C(N[C@H](C=O)CCCC)[C@@H](NC([C@@H](NC(C)=O)CC(C)C)=O)CC(C)C
Molecular Weight (g/mol) 383.53
Formula Weight 383.53
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.