missing translation for 'onlineSavingsMsg'
Learn More

MG-101, MedChemExpress

Product Code. 16427566
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16427566

Brand: Medchem Express HY18964

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MG-101 (ALLN) is an inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. MG-101 induces apoptosis and inhibits tumor growth, it can be used for the research of colon cancer.

MG-101 (ALLN) is an inhibitor of cysteine proteases which inhibits calpain I, calpain II, cathepsin B and cathepsin L with Kis of 190, 220, 150 and 500 pM, respectively. MG-101 induces apoptosis and inhibits tumor growth, it can be used for the research of colon cancer.

Specifications

Chemical Name or Material MG-101
Physical Form Solid
CAS 110044-82-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H37N3O4
For Use With (Application) Cancer-Kinase/protease
Synonym Calpain inhibitor I Ac-LLnL-CHO ALLN
Solubility Information DMSO : 16.67 mg/mL (43.46 mM; Need ultrasonic)
SMILES O=C(N[C@H](C=O)CCCC)[C@@H](NC([C@@H](NC(C)=O)CC(C)C)=O)CC(C)C
Molecular Weight (g/mol) 383.53
Formula Weight 383.53
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.