Learn More
MFSD9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA563104
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FSTAIGRTCI TDLQLTVGGA QASGTLIGVG QSVTAVGRII APLLSGVAQ Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
MFSD9 gene ontology annotations related to this gene include transmembrane transport.Specifications
MFSD9 | |
Polyclonal | |
Unconjugated | |
MFSD9 | |
major facilitator superfamily domain containing 9; major facilitator superfamily domain-containing protein 9; MFSD9; PSEC0066 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NBP5 | |
MFSD9 | |
Recombinant protein corresponding to Human MFSD9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only