missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFSD6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5142870
This item is not returnable.
View return policy
Description
Additional Information: The immuniznig peptide is thought to represent an extracellular domain. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
MFSD6 Polyclonal antibody specifically detects MFSD6 in Human samples. It is validated for Western Blot.
Specifications
| MFSD6 | |
| Polyclonal | |
| Unconjugated | |
| MFSD6 | |
| hMMR2; Macrophage MHC class I receptor 2 homolog; macrophage MHC receptor 2; major facilitator superfamily domain containing 6; major facilitator superfamily domain-containing protein 6; MFSD6; MMR2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 54842 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q6ZSS7 | |
| MFSD6 | |
| Peptide with sequence C-RPTTHPTNASHQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction