missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFSD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549207
This item is not returnable.
View return policy
Description
MFSD4 Polyclonal Antibody for Western Blot
MFSD4 gene ontology (GO) annotation includes glucose transmembrane transporter activity.
Specifications
| MFSD4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8N468 | |
| MFSD4A | |
| KLH conjugated synthetic peptide between 262-290 amino acids from the Central region of human MFSD4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| MFSD4A | |
| major facilitator superfamily domain containing 4; major facilitator superfamily domain containing 4A; major facilitator superfamily domain-containing protein 4; Major facilitator superfamily domain-containing protein 4A; MFSD4; MFSD4A; UNQ3064; UNQ3064/PRO9894 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 148808 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction