missing translation for 'onlineSavingsMsg'
Learn More

MFSD1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18427625 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18427625 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18427625 Supplier Novus Biologicals Supplier No. NBP176488AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MFSD1 Polyclonal antibody specifically detects MFSD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFSD1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias FLJ14153, major facilitator superfamily domain containing 1, major facilitator superfamily domain-containing protein 1, SMAP4, SMAP-4, Smooth muscle cell-associated protein 4, UG0581B09
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human MFSD1. The immunogen is located within amino acids 220 - 270 of MFSD1. Amino Acid Squence: LDQRAERILHKEQGKTGE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64747
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.