missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFRP Monoclonal Antibody (291204)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523996
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -4, -7, -8, -9, recombinant human Frizzled-5, -6, -10, or rmMFRP is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
MFRP is a member of the frizzled-related proteins. It may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen.
Specifications
| MFRP | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9BY79 | |
| MFRP | |
| Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| 291204 | |
| Unconjugated | |
| MFRP | |
| C1QTNF5; DKFZp586B0621; FLJ30570; MCOP5; Membrane frizzled-related protein; membrane-type frizzled-related protein; MFRP; MGC32938; NNO2; rd6 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 83552 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction