missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies mFreSR™
Description
- mFreSR™ is a serum-free cryopreservation medium designed for the cryopreservation of human embryonic and induced pluripotent stem cells (ES and iPS cells)
- mFreSR™ contains DMSO and is complete and ready-to-use
- Together with mTeSR™1, TeSR™2, or mTeSR™ Plus, mFreSR™ eliminates the use of feeders and serum
- Human ES and iPS cells cryopreserved in mFreSR™ have thawing efficiencies higher than reported conventional thawing methods using serum
Specifications
Specifications
| Cell Type | Pluripotent Stem Cells |
| For Use With (Application) | Cryopreservation |
| Quantity | 50 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Vous avez repéré une opportunité d'amélioration ?