missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFNG Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547834
This item is not returnable.
View return policy
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene is a member of the fringe gene family which also includes radical and lunatic fringe genes. They all encode evolutionarily conserved secreted proteins that act in the Notch receptor pathway to demarcate boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta-1,3-N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling.
Specifications
| MFNG | |
| Polyclonal | |
| Unconjugated | |
| MFNG | |
| AW546563; beta-1,3-N-acetylglucosaminyltransferase manic fringe; EC 2.4.1.222; manic fringe homolog; Mfng; MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 4242 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O00587 | |
| MFNG | |
| Mouse myeloma cell line NS0-derived recombinant human MFNG Gly37-Arg321. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction