Learn More
MFN2 Mouse anti-Human, Unconjugated, Clone: 5F3B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67487-1-IG-150UL
Description
The MFN2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MFN2. This antibody recognizes human, mouse, rat antigen. The MFN2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| MFN2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry | |
| 5F3B3 | |
| Unconjugated | |
| 150 μL | |
| 9927 | |
| IgG2a |