missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mfn2 Mouse anti-Human, Mouse, Rat, APC, Clone:S153-5, Abnova™
Mouse monoclonal antibody raised against partial recombinant human Mfn2.
Brand: Abnova MAB17081.100ug
This item is not returnable.
View return policy
Specifications
| Mfn2 | |
| Monoclonal | |
| APC | |
| Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
|
| Mfn2 | |
| Mouse | |
| 100 μg | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| S153-5 | |
| mitofusin 2 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide) | |
| D630023P19Rik, Fzo | |
| Recombinant protein corresponding to amino acids 370-600 at N-terminus of mouse Mfn2. | |
| 170731 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction