missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFN2, Mouse anti-Human, Clone: 6A8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009927-M01J.100ug
This item is not returnable.
View return policy
Description
Sequence: FKRQFVEHASEKLQLVISYTGSNCSHQVQQELSGTFAHLCQQVDVTRENLEQEIAAMNKKIEVLDSLQSKAKLLRNKAGWLDSELNMFTHQYLQPSRSpecifications
| MFN2 | |
| Monoclonal | |
| Unconjugated | |
| mitofusin 2 | |
| CMT2A/CMT2A2/CPRP1/HSG/KIAA0214/MARF | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9927 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6A8 | |
| PBS with no preservative; pH 7.4 | |
| NM_014874 | |
| MFN2 | |
| MFN2 (NP_055689, 661 a.a. ∼ 757 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction