missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFN1, Mouse anti-Human, Clone: 3C9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00055669-M04J.100ug
This item is not returnable.
View return policy
Description
Sequence: MAEPVSPLKHFVLAKKAITAIFDQLLEFVTEGSHFVEATYKNPELDRIATEDDLVEMQGYKDKLSIIGEVLSRRHMKVAFFGRTSSGKSSVINAMLWDKVLPSGIGHITNCFLSVEGTDGDKAYLMTEGSDEKKSVKTVNQLAHALHMDKDLKAGCLVRVFWPKAKCALLRDDLVLVDSPGTDVTTELDSWIDKFCLDADVFVLVANSESTLMNTEKHFFHKVNERLSKPNIFILNNRWDASASEPEYMEDVRRQHMERCLHFLVEELKVVNALEAQNRIFFVSAKEVLSARKQKAQGMPESGVALAEGFHARLQEFQNFEQIFEECISQSAVKTKFEQHTIRAKQILATVKNIMDSVNLAAEDKRHYSVEEREDQIDRLDFIRNQMNLLTLDVKKKIKEVTEEVANKVSCAMTDEICRLSVLVDEFCSEFHPNPDVLKIYKSELNKHIEDGMGRNLADRCTDEVNALVLQTQQEIIENLKPLLPAGIQDKLHTLIPCKKFDLSYNLNYHKLCSDFQEDIVFRFSLGWSSLVHRFLGPRNAQRVLLGLSEPIFQLPRSLASTPTAPTTPATPDNASQEELMITLVTGLASVTSRTSMGIIIVGGVIWKTIGWKLLSVSLTMYGALYLYERLSWTTHAKERAFKQQFVNYATEKLRMIVSSTSANCSHQVKQQIATTFARLCQQVDITQKQLEEEIARLPKEIDQLEKIQNNSKLLRNKAVQLENELENFTKQFLPSSNEESSpecifications
| MFN1 | |
| Monoclonal | |
| Unconjugated | |
| mitofusin 1 | |
| DKFZp762F247/FLJ20693/MGC41806/hfzo1/hfzo2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55669 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3C9 | |
| PBS with no preservative; pH 7.4 | |
| BC040557 | |
| MFN1 | |
| MFN1 (AAH40557, 1 a.a. ∼ 741 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction