missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10428-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.Specifications
| MFI2 | |
| Polyclonal | |
| Unconjugated | |
| MELTF | |
| CD228, MAP97, Melanotransferrin, MFI2, MTF1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288038, 30060, 4241 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P08582, Q9R0R1 | |
| Meltf | |
| MFI2 Fusion Protein Ag0700 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction