missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MFI2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552127
This item is not returnable.
View return policy
Description
Immunogen sequence: RSEDYELLCP NGARAEVSQF AACNLAQIPP HAVMVRPDTN IFTVYGLLDK AQDLFGDDHN KNGFKMFDSS NYHGQDLLFK DATVRAVPVG EKTTYRGWLG LDYVAALEGM SSQQC Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
The protein encoded by this gene is a cell-surface glycoprotein found on melanoma cells. The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.
Specifications
| MFI2 | |
| Polyclonal | |
| Unconjugated | |
| MELTF | |
| antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5; CD228; MAP97; Melanoma-associated antigen p97; melanoma-associated antigen p97, isoform 2; melanotransferrin; MELTF; membrane-bound transferrin-like protein; membrane-bound transferrin-like protein p97; Mfi2; Mtf; MTF1; p97 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4241 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P08582 | |
| MELTF | |
| Recombinant protein corresponding to Human MFI2. Recombinant protein control fragment (Product #RP-102453). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction