missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFGE8 (Lactadherin) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA582036
This item is not returnable.
View return policy
Description
Immunogen sequence: LKNNSIPDKQ ITASSSYKTW GLHLFSWNPS YARLDKQGNF NAWVAGSYGN DQWLQVDLGS SKEVTGIITQ GARNFGSVQF VASYKVAYSN DSANWTEYQD PRTGSSKIFP GNWDNHSHKK NLFETPILAR YVRILPVAWH NRIALRLEL
Reacts with 40-45 kDa glycoprotein in human milk fat globule membrane preparation.
Specifications
| MFGE8 (Lactadherin) | |
| Polyclonal | |
| Unconjugated | |
| MFGE8 | |
| BA46, EDIL1, hP47, HsT19888, MFG-E8, OAcGD3S, SED1, SPAG10 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 4240 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q08431 | |
| MFGE8 | |
| Recombit protein corresponding to Human MFGE8 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction