missing translation for 'onlineSavingsMsg'
Learn More

MFG Tray Chemical-Resistant Multipurpose Trays and Pans

Product Code. p-65064 Shop All Cole-Parmer Products
Click to view available options
Dimensions (L x W x H):
18 x 14 x 1 in.
Type:
Multi-purpose Tray
Unit Size:
Each

Product Code. 13074897

Brand: MFG Tray 0671582

Only null left
Add to basket
Edge
Add to basket

Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills

Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills

  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned

Order Info

Available in 1 in. deep (2.5cm) trays, shallow pans from 1 1/4 to 2 3/4 in. deep (3 to 7cm), and ventilated stacking pans from 2 3/4 to 4 1/8 in. deep (7 to 10.5cm).

Specifications

Dimensions (L x W x H) 18 x 14 x 1 in.
Color White
Height (Metric) 2.5 cm
Length (Metric) 46 cm
Material Fiberglass
Type Multi-purpose Tray
Width (Metric) 35.5 cm
For Use With (Application) Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Length (English) 18 in.
Height (English) 1 in.
Shape Rectangular
Temperature (English) -60°F to +250°F
Temperature (Metric) -51°C to +121°C
Width (English) 14 in.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.