missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MFG-E8 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67797-1-IG-20UL
This item is not returnable.
View return policy
Description
The MFG-E8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MFG-E8. This antibody recognizes human antigen. The MFG-E8 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| MFG-E8 | |
| Monoclonal | |
| 1000 μg/mL | |
| BA46; EDIL1; HsT19888; MFG-E8; OAcGD3S; SED1; SPAG10; hP47 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 43 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A1A9 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 4240 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction