Learn More
MFG-E8 Antibody (MFG-06), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44410-0.1mg
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Monoclonal | |
| MFGE8 | |
| IgG1 κ |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 1 - 2 ug/ml | |
| Store at 4C. |