missing translation for 'onlineSavingsMsg'
Learn More

MFF Rabbit anti-Human, Mouse, Bovine, Polyclonal, Novus Biologicals™

Product Code. 18393542
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393542 25 μg 25µL
18318241 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18393542 Supplier Bio-Techne Supplier No. NBP31194725UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MFF Polyclonal antibody specifically detects MFF in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFF
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200
Formulation Neat Serum
Gene Alias mitochondrial fission factor
Host Species Rabbit
Immunogen Human recombinant protein. (UniProt: Q9GZY8)
Purification Method Unpurified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56947
Target Species Human, Mouse, Bovine
Content And Storage Store at -20C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.