missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MFAP5 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552706

Product Code. 15700661

  • £446.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GVNSQRGDDV TQATPETFTE DPNLVNDPAT DETVLAVLAD IAPSTDDLAS LSEKNTTAEC WDEKFTCTRL YSVHRPVKQC IHQLCFTSLR RMYIVNKEIC SRLVCKEHEA MKDELCRQMA GLPPRRLRRS NYFRLPPCEN V Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.

This gene encodes a 25-kDa microfibril-associated glycoprotein which is rich in serine and threonine residues. It lacks a hydrophobic carboxyl terminus and proline-, glutamine-, and tyrosine-rich regions, which are characteristics of a related 31- kDa microfibril-associated glycoprotein (MFAP2). The close similarity between these two proteins is confined to a central region of 60 aa where precise alignment of 7 cysteine residues occurs. The structural differences suggest that this encoded protein has some functions that are distinct from those of MFAP2.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MFAP5
Polyclonal
Unconjugated
MFAP5
AAT9; MAGP2; MAGP-2; MFAP5; MFAP-5; Microfibril-associated glycoprotein 2; microfibril-associated glycoprotein-2; microfibrillar associated protein 5; microfibrillar-associated protein 5; MP25; THE1A-MFAP5
Rabbit
Antigen affinity chromatography
RUO
8076
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.20 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q13361
MFAP5
Recombinant protein corresponding to Human MFAP5. Recombinant protein control fragment (Product #RP-89743).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.