missing translation for 'onlineSavingsMsg'
Learn More

MFAP3L Antibody (1059107) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30669884 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30669884 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30669884 Supplier Novus Biologicals Supplier No. FAB11325JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MFAP3L Monoclonal antibody specifically detects MFAP3L in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen MFAP3L
Applications Immunocytochemistry
Classification Monoclonal
Clone 1059107
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias HSD39, HSD-39, microfibrillar-associated protein 3-like, NYD-sp9, testis development protein NYD-SP9
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human MFAP3L, Met1-Met149, Accession # O75121
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9848
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.