missing translation for 'onlineSavingsMsg'
Learn More

MF63, MedChemExpress

Product Code. 16465618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465618 10 mg 10mg
16451787 50 mg 50mg
16428317 5 mg 5mg
16426156 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16465618 Supplier Medchem Express Supplier No. HY13283
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress MF63 is a selective mPGES-1 inhibitor with an IC50 of 0.9 nM and 1.3 nM for pig mPGES-1 and human mPGES-1 enzyme, respectively.

MF63 is a selective mPGES-1 inhibitor with an IC50 of 0.9 nM and 1.3 nM for pig mPGES-1 and human mPGES-1 enzyme, respectively.

Specifications

Chemical Name or Material MF63
CAS 892549-43-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H11ClN4
Quantity 10 mg
Solubility Information DMSO : ≥ 43 mg/mL (113.51 mM)
SMILES ClC1=CC(C2=C3C=CC=C2)=C(C4=C3N=C(C5=C(C=CC=C5C#N)C#N)N4)C=C1
Molecular Weight (g/mol) 378.81
Formula Weight 378.81
Percent Purity 99.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.