missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Mixed Cellulose Ester Membranes, Triton-free

Composed of mixed cellulose esters (nitrate and acetate)

Brand:  Merck GSTF02500

Code : 71

Additional Details : CAS Number : 9004-70-0 Weight : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II


 View more versions of this product

Product Code. 10309610

  • £129.00 / Pack of 100
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Composed of mixed cellulose esters (nitrate and acetate)

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications. MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.Features & Benefits: - Versatile filter for biological and environmental monitoring applications - Available in a range of pore sizes, colored black or white, with or without a gridded surface - Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications

Specifications

Not Autoclavable
Mixed Cellulose Esters (MCE) (Nitrate and Acetate); Triton-Free
Non-sterile
Clarification of fluids; Sterilizing filtration of tissue culture media, additives, antibiotics; Air monitoring; Particulate analysis; Particulate removal; Biological assays (receptor binding studies, TCA precipitates, etc.).
Filter Membranes
25 mm
White
0.22 μm
18 mL/min x cm2
Plain
Hydrophilic
25 mm
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MCE TRITON FREE .22UM WH PL 25MM 100/PK

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use to extinguish.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers