missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, 0.8 μm

Product Code. 10442321 Shop All Merck Products
Change view
Click to view available options
Diameter (Metric):
13 mm
25 mm
37 mm
47 mm
90 mm
142 mm
Color:
Black
White
Type:
Filter Membranes
Liquid Monitor Refill
Matched-weight Filter Pairs
Unit Size:
Pack of 100
Pack of 50
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Color Type unitSize
10442321 47 mm White Filter Membranes Pack of 100
10024670 13 mm White Filter Membranes Pack of 100
10004560 25 mm Black Filter Membranes Pack of 100
10310131 25 mm White Filter Membranes Pack of 100
11576395 37 mm White Matched-weight Filter Pairs Pack of 50
10780303 37 mm White Liquid Monitor Refill Pack of 100
10278050 47 mm Black Filter Membranes Pack of 100
10298240 47 mm White Matched-weight Filter Pairs Pack of 50
10074690 90 mm White Filter Membranes Pack of 100
11586395 142 mm White Filter Membranes Pack of 50
10442131 37 mm White Filter Membranes Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10442321 Supplier Merck Supplier No. AAWP04700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mixed cellulose esters (MCE) membrane, hydrophilic

Designed for air monitoring, bioassays, dairy microbiology, particle monitoring and particle removal. MF-Millipore™ Mixed Cellulose Ester Membranes, 0.80μm pore size are available in a range of diameters. Composed of mixed cellulose membrane in black or white with a plain or gridded surface.

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.

Features & Benefits:

  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bubble Point ≥1.0 bar, air with water
Color White
Filter Type Membrane Filter
Material MCE
Pore Size 0.8 μm
Sterility Non-sterile
Flow Rate 190 mL/min x cm2
For Use With (Application) For air monitoring, bioassays, dairy microbiology, particle monitoring, and particle removal
Gas Flow Rate 16 L/min/ cm2
Grade AAWP
Hydrophobicity Hydrophilic
No. per Case 100Pack
Quantity 100/Pk.
Surface Type Plain
Thickness (Metric) 150 μm
Type Filter Membranes
Wettability Hydrophilic
Diameter (Metric) 47 mm
Show More Show Less
compliance-icons
Product Identifier
  • MCE .8UM WH PL 47MM 100PK
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.
Supplemental information
  • MIXTURE LIST-Contain: nitrocellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.