missing translation for 'onlineSavingsMsg'
Learn More

Mexiletine hydrochloride

Product Code. p-7077252 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15688685

Brand: Thermo Scientific Acros 372910050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5370-01-4 | C11H17NO·HCl | 215.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-4-5370
Molecular Formula C11H17NO·HCl
Molecular Weight (g/mol) 215.72
InChI Key NFEIBWMZVIVJLQ-UHFFFAOYNA-N
Synonym mexiletine hydrochloride, mexitil, mexiletine hcl, mexiletene hydrochloride, 1-2,6-dimethylphenoxy-2-propanamine hydrochloride, 1-2,6-dimethylphenoxy propan-2-amine hydrochloride, 2-propanamine, 1-2,6-dimethylphenoxy-, hydrochloride, mexitil tn, 1-2,6-xylyloxy-2-aminopropane hydrochloride
PubChem CID 21467
ChEBI CHEBI:6917
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
SMILES CC1=C(C(=CC=C1)C)OCC(C)N.Cl

Specifications

Melting Point 202.0°C
Quantity 5 g
Infrared Spectrum Authentic
Linear Formula (CH3)2C6H3OCH2CH(NH2)CH3·HCl
Merck Index 15,6248
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 215.72
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Mexiletine hydrochloride
compliance-icons
Product Identifier
  • Mexiletine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Methyl-2-(2,6-xylyloxy)ethylammonium chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.