missing translation for 'onlineSavingsMsg'
Learn More

Mexiletine hydrochloride

Product Code. 15698685 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15698685 25 g 25g
15688685 5 g 5g
15608695 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15698685 Supplier Thermo Scientific Acros Supplier No. 372910250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 5370-01-4 | C11H17NO·HCl | 215.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-4-5370
Molecular Formula C11H17NO·HCl
Molecular Weight (g/mol) 215.72
InChI Key NFEIBWMZVIVJLQ-UHFFFAOYNA-N
Synonym mexiletine hydrochloride, mexitil, mexiletine hcl, mexiletene hydrochloride, 1-2,6-dimethylphenoxy-2-propanamine hydrochloride, 1-2,6-dimethylphenoxy propan-2-amine hydrochloride, 2-propanamine, 1-2,6-dimethylphenoxy-, hydrochloride, mexitil tn, 1-2,6-xylyloxy-2-aminopropane hydrochloride
PubChem CID 21467
ChEBI CHEBI:6917
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
SMILES CC1=C(C(=CC=C1)C)OCC(C)N.Cl

Specifications

Melting Point 202.0°C
Quantity 25 g
Infrared Spectrum Authentic
Linear Formula (CH3)2C6H3OCH2CH(NH2)CH3·HCl
Merck Index 15,6248
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 215.72
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Mexiletine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.