missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MEX3D Polyclonal antibody specifically detects MEX3D in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | MEX3D |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 350 |
| Dilution | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | KIAA2031Tino, mex-3 homolog D (C. elegans), OK/SW-cl.4, ring finger (C3HC4 type) and KH domain containing 1, ring finger and KH domain containing 1, RING finger and KH domain-containing protein 1, RING finger protein 193, RKHD1MEX3, RNA-binding protein MEX3D, RNF193MEX-3D, TINObcl-2 ARE RNA binding protein |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Rkhd1. The immunogen is located within amino acids 360 - 410 of Rkhd1. Amino Acid Squence: WAKTPNQGRRPPTAT |
| Purification Method | Peptide affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?