missing translation for 'onlineSavingsMsg'
Learn More

MEX3D Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18429492 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429492 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429492 Supplier Novus Biologicals Supplier No. NBP177332AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEX3D Polyclonal antibody specifically detects MEX3D in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEX3D
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias KIAA2031Tino, mex-3 homolog D (C. elegans), OK/SW-cl.4, ring finger (C3HC4 type) and KH domain containing 1, ring finger and KH domain containing 1, RING finger and KH domain-containing protein 1, RING finger protein 193, RKHD1MEX3, RNA-binding protein MEX3D, RNF193MEX-3D, TINObcl-2 ARE RNA binding protein
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Rkhd1. The immunogen is located within amino acids 360 - 410 of Rkhd1. Amino Acid Squence: WAKTPNQGRRPPTAT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 399664
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.