missing translation for 'onlineSavingsMsg'
Learn More

Mevastatin, 98%

Product Code. 15493959
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493959 25 mg 25mg
15473959 5 mg 5mg
15483959 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493959 Supplier Thermo Scientific Alfa Aesar Supplier No. J61357.MB
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 73573-88-3 | C23H34O5 | 390.52 g/mol

Mevastatin (Compactin) inhibits HMGCR (HMG-CoA reductase) , induces apoptosis which in turn inhibits isoprenoid biosynthesis and therefore blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Mevastatin causes cells to arrest early in the G1 phase. Mevastatin is a close structural analog of lovastatin and both agents have the same biochemical and pharmacological activities. Mevastatin is inactive in cell-free assays. In cells however, it is hydrolyzed to the active free acid form by intracellular esterases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mevastatin (Compactin) inhibits HMGCR (HMG-CoA reductase) , induces apoptosis which in turn inhibits isoprenoid biosynthesis and therefore blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Mevastatin causes cells to arrest early in the G1 phase. Mevastatin is a close structural analog of lovastatin and both agents have the same biochemical and pharmacological activities. Mevastatin is inactive in cell-free assays. In cells however, it is hydrolyzed to the active free acid form by intracellular esterases.

Solubility
Soluble in ethanol at 25 mg/mL and in DMSO at 20mg/ml

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73573-88-3
Molecular Formula C23H34O5
Molecular Weight (g/mol) 390.52
MDL Number MFCD05662341
InChI Key AJLFOPYRIVGYMJ-INTXDZFKSA-N
Synonym mevastatin, compactin, mevastatinum, mevastatina, mevastatine, mevastatinum inn-latin, antibiotic ml 236b, mevastatin inn, compactin penicillium, mevastatine inn-french
PubChem CID 64715
ChEBI CHEBI:34848
IUPAC Name [(1S,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1CCC=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Color White
Quantity 25 mg
Beilstein 1269441
Merck Index 14,6164
Solubility Information Soluble in ethanol at 25 mg/mL and in DMSO at 20mg/ml
Formula Weight 390.52
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Mevastatin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.