missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Mevastatin

Product Code. 16493337
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16493337 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16493337 Supplier MedChemExpress Supplier No. HY17408
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mevastatin (Compactin) is a first HMG-CoA reductase inhibitor that belongs to the statins class. Mevastatin is a lipid-lowering agent, and induces apoptosis, arrests cancer cells in G0/G1 phase. Mevastatin also increases endothelial nitric oxide synthase (eNOS) mRNA and protein levels. Mevastatin has antitumor activity and has the potential for cardiovascular diseases treatment.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 390.51 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 73573-88-3
Product Type Natural Products
Purity 98.45%
Solubility DMSO : 250 mg/mL (640.19 mM∣Need ultrasonic)
Synonym Compactin∣ ML236B
Target HMG-CoA Reductase (HMGCR)∣Bacterial∣Autophagy∣Apoptosis∣Antibiotic
Quantity 50 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.