missing translation for 'onlineSavingsMsg'
Learn More

(±)-Mevalonolactone, 97%

Product Code. 10119699 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10119699 5 g Glass Bottle 5g
10422790 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10119699 Supplier Thermo Scientific Acros Supplier No. 428300050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 674-26-0 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 674-26-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00006648
InChI Key JYVXNLLUYHCIIH-UHFFFAOYNA-N
Synonym mevalonolactone, dl-mevalonic acid lactone, dl-mevalonolactone, 4-hydroxy-4-methyltetrahydro-2h-pyran-2-one, mevalonic acid lactone, mevalonic lactone, dl-mevalolactone, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl-, .+/-., 1-tetrahydro-4-hydroxy-4-methyl-2h-pyran-2-one
PubChem CID 10428
IUPAC Name 4-hydroxy-4-methyloxan-2-one
SMILES CC1(O)CCOC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 28.0°C
Boiling Point 145.0°C to 150.0°C (5.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 5 g
Solubility Information Solubility in water: soluble
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material (±)-Mevalonolactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.