missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metyrapone, MedChemExpress
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Metyrapone inhibits synthesis of endogenous adrenal corticosteroid, decreases glucocorticoid levels, and also affects behavior and emotion. In addition, Metyrapone increases the efficiency of autophagic process via downregulation of mTOR pathway, and interacts with Pseudomonas putida cytochrome P-450. Metyrapone can be used for researching Cushing's syndrome and depression.
Specifications
Specifications
| Chemical Name or Material | Metyrapone |
| CAS | 54-36-4 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C14H14N2O |
| Quantity | 500 mg |
| Synonym | Su-4885 |
| Solubility Information | DMSO : 100 mg/mL (441.95 mM; Need ultrasonic) ∣H2O : ≥ 38 mg/mL (167.94 mM) |
| SMILES | CC(C1=CC=CN=C1)(C)C(C2=CC=CN=C2)=O |
| Molecular Weight (g/mol) | 226.27 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction