Learn More
Mettler Toledo™ Titration Burette For Titration Sensor
Product Code. 10385463 Shop All Mettler Toledo Products
Change view
Click to view available options
Volume (Metric):
20 mL
Unit Size:
Each
Product Code. Volume (Metric) Unit Size
10385463 20 mL Each
1 options
Product Code. 10385463 Supplier Mettler Toledo™ Supplier No. 51107502
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Includes
RFID chip
Features optional ceramic valve discs for crystallizing titrants.
  • Allow rapid titrant change
  • Red protection tubing against light
  • Replaceable PTFE valve discs
  • PTFE pistons w/tripple lip seal assure good tightness
Compliance
ISO 8655-3
TRUSTED_SUSTAINABILITY
Specifications
Type Titration Burette
Includes RFID chip
Volume (Metric) 20 mL