missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Hydrofluoric pH Electrode

Product Code. 10523185 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
S7
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connector Types unitSize
10523185 S7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10523185 Supplier METTLER TOLEDO™ Supplier No. 51343176
Add to basket
Add to basket
This item is not returnable. View return policy

A pH electrode featuring a glass body that is resistant to hydrofluoric acid.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Hydrofluoric (HF) bearing sample

Accessories & Care

  • Ceramic junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Description pH electrode InLab Hydrofluoric
Connector Types S7
Junction Ceramic
Material Glass
pH Range 1 to 11
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) Improves service life and measurement accuracy at differing sample temperatures
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Membrane HF Glass
Model InLab™ Hydrofluoric
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Tip Type Bulb
Length (English) 4.72 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.