Learn More
Mettler Toledo™ RS-P42 Dot Matrix Printer
For use with Excellence XS and Excellence Plus XP balances. Mettler Toledo RS-P42 Dot Matrix Printer meets GLP/GMP/ISO requirements and avoids transcription errors and ensure complete data records.
Brand:  Mettler Toledo™ 00229265
Product Code. 10563572
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Uses normal and adhesive paper
  • Prints weighing results, regular adjustments and—depending on the balance—individual configuration application results
  • Speed: 24 characters per line, 1 line per second
  • Dimensions: 8-1/4L × 6-1/8W × 3-3/8 in.H (21 × 15.7× 8.5cm)
  • Net weight: 3-3/16 lb. (1.5kg)
  • For 115-230V 50/60Hz