Learn More
Mettler Toledo™ ST24 Septum Stopper for V20S Titrator
Product Code. 13172162 Shop All Mettler Toledo Products
Change view
Click to view available options
Unit Size:
Pack of 10
Product Code. unitSize
13172162 Pack of 10
1 options
Product Code. 13172162 Supplier Mettler Toledo™ Supplier No. 00023950
Only null left
Add to basket
Add to basket
Septum stopper used to seal the titration vessel on METTLER TOLEDO Volumetric Karl Fischer titrators.
  • NS24
  • Standard taper 24
TRUSTED_SUSTAINABILITY
Specifications
Type Septa
For Use With (Application) Water content determination