missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- NMR tubes
- Small volume samples
- Small measuring vessels
Accessories & Care
- Ceramic junction
- Refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Order Info
Metal Half-Cells (01-920-162, -163 and -164) and U-Electrodes (09-320-200 and -201) also available separately.
Specifications
Specifications
| Description | pH electrode InLab NMR |
| Connector Types | S7 |
| Junction | Ceramic |
| Material | Glass |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 80°C |
| Type | Combination Electrode |
| Filling Solution Type | 3mol/L KCl AgCl saturated |
| For Use With (Application) | For particularly long test tubes, vials or vessels |
| For Use With (Equipment) | S7 to BNC (30281915) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?