missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Redox ORP Electrodes

An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.

Brand:  METTLER TOLEDO™ 51343205

Code : 59

Additional Details : Weight : 0.07000kg

 View more versions of this product

Product Code. 10683004

  • £306.00 / Each
Estimated Shipment: 10-07-2024
to see stock.
Add to basket

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Measuring oxidation/reduction potential (ORP)
  • Drinking water
  • Cooling towers
  • Swimming pools
  • Potable water supplies
  • Waste water
  • General lab

Accessories & Care

  • Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications

Specifications

S7
Glass
Combination Electrode; Silver ring indicator
For measuring the oxidation/reduction power of solution
ARGENTHAL with Ag+ trap
1mol/L KNO3
0°C to 100°C
120 mm
12 mm
Ceramic
0°C to 100°C
1mol/L KNO3
S7 to BNC (30281915)
InLab™ Redox Ag
32°F to 212°F
4.72 in.
0.47 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers