missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Redox ORP Electrodes

Product Code. 10353713 Shop All Mettler Toledo Products
Change view
Click to view available options
Model:
InLab Redox
InLab Redox Ag
InLab Redox Ag805
InLab Redox Au
InLab Redox Flow
InLab Redox Micro
InLab Redox Pt805
InLab Redox-L
Type:
Combination Electrode
Half Cell Electrode
Electrode:
Gold Ring Indicator
Platinum Ring Indicator
Silver Ring Indicator
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Type Electrode unitSize
10353713 InLab Redox-L Combination Electrode Platinum Ring Indicator Each
10786733 InLab Redox Combination Electrode Platinum Ring Indicator Each
10683004 InLab Redox Ag Combination Electrode Silver Ring Indicator Each
10735583 InLab Redox Au Combination Electrode Gold Ring Indicator Each
10028142 InLab Redox Flow Combination Electrode Platinum Ring Indicator Each
10531645 InLab Redox Micro Combination Electrode Platinum Ring Indicator Each
10732722 InLab Redox Pt805 Half Cell Electrode Platinum Ring Indicator Each
10748972 InLab Redox Ag805 Half Cell Electrode Silver Ring Indicator Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10353713 Supplier METTLER TOLEDO™ Supplier No. 51343202
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

An ORP/Redox electrode featuring a metal ring indicator designed for measuring oxidation/reduction potential; ideal for water and other general applications.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Measuring oxidation/reduction potential (ORP)
  • Drinking water
  • Cooling towers
  • Swimming pools
  • Potable water supplies
  • Waste water
  • General lab

Accessories & Care

  • Ceramic junction (excluding the InLab Redox Flow with movable glass sleeve junction)
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • InLab Redox Pt805 and InLab Redox Ag805: no electrolyte required and must be paired with InLab Reference (51343190)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Spezifikation

Connector Types S7
Junction Ceramic
Material Glass
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Electrode Platinum Ring Indicator
Filling Solution Type 3mol/L KCl
For Use With (Application) For measuring the oxidation/reduction power of solution
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Model InLab Redox-L
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 6.69 in.
Length (Metric) 170 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.