Learn More
Mettler Toledo™ LabX™ Direct Balance Software
Product Code. 12701261 Shop All Mettler Toledo Products
Change view
Click to view available options
LabX Direct Balance Software
Product Code.
12701261 LabX Direct Balance Software
1 options
Product Code. 12701261 Supplier Mettler Toledo™ Supplier No. 11120340
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Increases the efficiency and accuracy of weighing applications. Fisher Scientific - Mettler Toledo LabX Direct Balance Software is an easy-to-use software designed to transfer data from one balance into an open application like an Excel™ spreadsheet.
  • Control of all Mettler Toledo balances equipped with an RS-232 interface
  • Easy setup
  • Add time and date to your data
  • Data lay-out configurable
  • Transfer data into any Windows™-based program like Excel
  • Support of barcode reader
  • User defined information can be automatically added to data
  • User manual available as pdf file
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Balance
For Use With (Application) Increases the efficiency and accuracy of weighing
Type Direct Balance Software