Learn More
Mettler Toledo™ LabX™ Balance Express Software
Shop All Mettler Toledo Products
Click to view available options
For Use With (Equipment):
Excellence Series Balance
Includes:
Balance Express Starter Pack: For installation on
Description
- Express edition
- Connect one instrument with option to up to 3 balances
- Graphical method editor supports any weighing application
- Built in method library for quick weighing method development
- Comprehensive and customizable report editor
LabX seamlessly integrates with third-party systems such as LIMS, ELN, SAP and CDS
Specifications
Specifications
| For Use With (Equipment) | Excellence Series Balance |
| For Use With (Application) | Weighs different classes of material with improved |
| Includes | Balance Express Starter Pack: For installation on |
| Type | Balance Software |