Learn More
Mettler Toledo™ LabX™ Balance Express Software
Product Code. 12767191 Shop All Mettler Toledo Products
Change view
Click to view available options
For Use With (Equipment):
Excellence Series Balance
Includes:
Balance Express Starter Pack: For installation on
Product Code. For Use With (Equipment) Includes
12767191 Excellence Series Balance Balance Express Starter Pack: For installation on
1 options
Product Code. 12767191 Supplier Mettler Toledo™ Supplier No. 11153120
Weighs different classes of material with improved documentation allowing electronic integration of weights and sample IDs with other software systems within lab.
  • Express edition
  • Connect one instrument with option to up to 3 balances
  • Graphical method editor supports any weighing application
  • Built in method library for quick weighing method development
  • Comprehensive and customizable report editor
LabX seamlessly integrates with third-party systems such as LIMS, ELN, SAP and CDS
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Excellence Series Balance
For Use With (Application) Weighs different classes of material with improved
Includes Balance Express Starter Pack: For installation on
Type Balance Software