missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Versatile pH Electrode

Product Code. 10690124 Shop All Mettler Toledo Products
Click to view available options
Connector Types:
BNC
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10690124

Brand: METTLER TOLEDO™ 51343031

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A plastic, refillable pH electrode with an integrated temperature sensor ideal for measuring liquids. Electrode features a replaceable ceramic junction, which offers a solution to contaminated junctions without the higher cost or cleaning efforts required with open or sleeve junctions.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Drinking water
  • Soft drinks
  • Beer
  • General lab

Accessories & Care

  • Replaceable ceramic junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types BNC
Junction Ceramic
Material Polysulfone
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode; Integrated Temperature Probe
Filling Solution Type 3mol/L KCl
For Use With (Application) Alternative for measurements in simple samples, offer an outstanding price/performance ratio
For Use With (Equipment) Fixed Cable to sensor
Internal Reference ARGENTHAL with Ag+ trap
Membrane U Glass
Model InLab™ Versatile Pro
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.72 in.
Length (English) Cable 0.84 in.
Length (Metric) 120 mm
Length (Metric) Cable 1.2 m
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.