missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Halogen Moisture Analyzer Glass Fiber Discs

Product Code. 12773165 Shop All Mettler Toledo Products
Click to view available options
Diameter (Metric):
88.9 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12773165

Brand: Mettler Toledo™ 00214464

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Mettler Toledo filters are the only filters that are officially supported by Mettler Toledo Moisture Analyzers for optimal use and guaranteed performance.

For use with samples with high moisture content. The absorbent glass fiber filter is beneficial for liquid or pasty substances. For a further reduction in analysis time.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color White
Disposable Single-use
Material Glass
Shape Round
Sterility Non-sterile
Type Moisture Disc
For Use With (Equipment) Mettler Toledo Halogen Moisture Analyzer
Diameter (Metric) 88.9 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.