Learn More
Mettler Toledo™ Electrode Cable for Karl Fischer™ Titrator
Product Code. 12815993 Shop All Mettler Toledo Products
Change view
Click to view available options
Connector Types:
Lemo
Unit Size:
Each
Product Code. Connector Types unitSize
12815993 Lemo Each
1 options
Product Code. 12815993 Supplier Mettler Toledo™ Supplier No. 00089602
Currently Unavailable: We have no information when this product will be available.
View alternative products
Accessory for use with V10/20S and G10/20S Compact Titrators.
  • Type: SC-Lemo
  • For use with standard and Plug and Play electrodes
  • For V10/20S (01-914-315/ 01-914-315) or G10/20S (01-914-318/ 01-914-319) series titrators
  • Length: 100cm
TRUSTED_SUSTAINABILITY
Specifications
Connector Types Lemo
For Use With (Equipment) Standard and Plug and Play Electrodes
For Use With (Application) Connection to the titrators
Includes S12 grey connection
Length (Metric) Cable 100 cm