missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Interchangeable Buret, for Mettler Toledo™ Titrators

Product Code. 10529290 Shop All Mettler Toledo Products
Change view
Click to view available options
Volume (Metric):
5 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10529290 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10529290 Supplier Mettler Toledo™ Supplier No. 51107500
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with V20 Karl Fischer and G20 Compact Titrator.

  • Volume: 5mL
TRUSTED_SUSTAINABILITY

Specifications

Type Titration Burette
Volume (Metric) 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.