Learn More
Description
Includes
AC adapter appropriate to national standard
For simple, easy, weighing tasks that need to be done frequently, but with assured accuracy: piece counting, percent weighing, ±weighing, dynamic weighing, and calculation with a custom “free factor”
Key Features
- Logical menu guidance with only a few keys for full function
- Portable; battery operation operates for 25 to 30 hours
- Automatic shutoff saves battery life
- Built-in overload protection
- Specific gravity and density measurements with built-in below-balance weighing mechanism
- Weighing modes: g, kg, mg, carats, lb., oz., ounces troy, grains, pennyweight, mommes, mesghals, taels (three variants), ticals, tolas and bahts
- Large, high-contrast, seven-segment LCD
- Automatic calibration with external weights
- Large weighing pan supports loading efficiency
- PL-S models offer the option of a built-in rechargeable storage battery
Order Info
Electrical Requirements: 105/120V 50/60Hz. Dimensions: 8.87 x 7.62 x 5.37 in. (22.5 x 19.4 x 13.7cm); except PL303 and PL3002 which are 13.25 x 9.37 x 11.25 (33.5 x 23.8 x 28.7cm). Net Weight: 3.37 lb. (1.5kg); except PL83-S and PL303 which are 12.12 lb. (5.5kg). A variety of accessories are available including optional carrying case, in-use covers, and power adapters.
Warranty
One year
Specifications
Specifications
| Capacity | 610 g |
| Readability | 0.01 g |
| Model | PL602-S/03 |
| Stabilization Time | 2.5 sec. |
| Type | Electronic Toploading Balance |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.