Learn More
Mettler Toledo™ Balancelink™ Software
Product Code. 12714772 Shop All Mettler Toledo Products
Click to view available options
:
Balancelink
Product Code. 12714772
Brand: Mettler Toledo™ 00237010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Requires
MS Windows 2000™(SP4 or higher)/ MS Windows XP™ (SP1 or higher), Intel Pentium II™ 450 MHz, 128 MB RAM
Facilitates direct transfer of measurement results into application program
  • Integration of highly diverse Mettler Toledo measuring instruments
  • Configurable interface for integration of other instruments
  • Filters and macros allow user-defined data format
  • Program settings can be saved for future use
  • Instrument specific configuration options can be protected and only accessed by authorised users
  • Password-protected settings
  • Date and time stamp for each measurement result (GLP/GMP)
  • Data can be written automatically into a simple text document
  • Automatic conversion of measurement values
  • Supports five languages: English, French, German, Italian and Spanish
  • Allows data communication between awide range of Mettler Toledo instruments and application program such as MS Word™, Excel™, MS Access™ and many more
  • Online help also available
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Balance
For Use With (Application) Transfers data into desktop applications, integrat
Type Balancelink™ Software