missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Balance Printer Accessories: Printer Paper
Printer paper for use with Mettler Toledo™ balances.
Brand: Mettler Toledo™ 72456
This item is not returnable.
View return policy
Description
- Color: White
- Five rolls per pack
- For use with Mettler Toledo balances: LC-P45, LC-P43, HA-P43, HR-73P
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?